Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531022 Supplier Invitrogen™ Supplier No. PA531022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Raji. Predicted reactivity: Mouse (80%), Rat (80%), Dog (85%), Pig (84%), Rabbit (87%), Rhesus Monkey (98%), Chimpanzee (99%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but some of the full length sequences are not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ASB3
Gene Accession No. Q9Y575
Gene Alias 2400011J03Rik; ankyrin repeat and SOCS box containing 3; ankyrin repeat and SOCS box protein 3; ankyrin repeat and SOCS box-containing 3; ankyrin repeat and SOCS box-containing 3-like; ankyrin repeat and SOCS box-containing protein 3; ankyrin repeat-containing SOCS box protein 3; ASB3; Asb-3; EAR1; FLJ10123; FLJ10421; HREV; MGC12531; MGC132002; MGC996; THRAL
Gene Symbols ASB3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ASB3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51130
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.