Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599098
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599098 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599098 Supplier Invitrogen™ Supplier No. PA599098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASB4 Polyclonal Antibody for Western Blot, ELISA

Probable substrate-recognition component of a SCF-like ECS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ASB4
Gene Accession No. Q9WV71, Q9Y574
Gene Alias 8430401O13Rik; ankyrin repeat and SOCS box containing 4; ankyrin repeat and SOCS box protein 4; ankyrin repeat and SOCS box-containing 4; ankyrin repeat and SOCS box-containing protein 4; ASB4; ASB-4; AV117194; AW060255
Gene Symbols ASB4
Host Species Rabbit
Immunogen Recombinant Human Ankyrin repeat and SOCS box protein 4 protein (166-321AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500017, 51666, 65255
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.