Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASB5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528535
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse muscle. Predicted reactivity: Mouse (92%), Rat (83%), Rabbit (94%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ASB5 encodes a protein that is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signaling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but their full length sequences are not known.
Specifications
ASB5 | |
Polyclonal | |
Unconjugated | |
ASB5 | |
1110018D09Rik; ankyrin repeat and SOCS box containing 5; ankyrin repeat and SOCS box protein 5; ankyrin repeat and SOCS box-containing 5; ankyrin repeat and SOCs box-containing protein 5; ASB5; ASB-5; SOCS box protein ASB-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140458, 76294 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8WWX0, Q9D1A4 | |
ASB5 | |
Recombinant fragment corresponding to a region within amino acids 48 and 329 of Human ASB5. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction