Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASB7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598848
Description
ASB7 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene belongs to a family of ankyrin repeat proteins that, along with four other protein families, contains a C-terminal SOCS box motif. Growing evidence suggests that the SOCS box acts as a bridge between specific substrate-binding domains and the more generic proteins that comprise a large family of E3 ubiquitin protein ligases. In this way, SOCS box containing proteins may regulate protein turnover by targeting proteins for polyubiquination and, therefore, for proteasome-mediated degradation. Two alternative transcripts encoding different isoforms have been described.
Specifications
ASB7 | |
Polyclonal | |
Unconjugated | |
ASB7 | |
AI449039; ankyrin repeat and SOCS box containing 7; ankyrin repeat and SOCS box protein 7; ankyrin repeat and SOCS box-containing 7; ankyrin repeat and SOCS box-containing protein 7; ASB7; Asb-7; D030055C23Rik | |
Rabbit | |
Protein G | |
RUO | |
140460 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9H672 | |
ASB7 | |
Recombinant Human Ankyrin repeat and SOCS box protein 7 protein (1-274AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction