Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASB8 Antibody (OTI3A12), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272400AF532
Description
ASB8 Monoclonal specifically detects ASB8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ASB8 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
ASB8 | |
Full length recombinant protein of human ASB8 (NP_077000) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI3A12 | |
Western Blot, Immunohistochemistry | |
ankyrin repeat and SOCS box containing 8, ankyrin repeat and SOCS box protein 8, ankyrin repeat and SOCS box-containing 8, ASB-8, FLJ21255, MGC5540 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
140461.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction