Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550354 Supplier Invitrogen™ Supplier No. PA550354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ASB8 protein.

May be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ASB8
Gene Accession No. Q9H765
Gene Alias 4930539L19Rik; AI788835; ankyrin repeat and SOCS box containing 8; ankyrin repeat and SOCS box protein 8; ankyrin repeat and SOCS box-containing 8; ankyrin repeat and SOCS box-containing protein 8; ASB8; ASB-8; C430011H06Rik; PP14212
Gene Symbols ASB8
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the N terminal of human ankyrin repeat and SOCS box containing 8.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140461
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.