Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASB8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551887
Description
Immunogen sequence: QSIQSKYSLS ERLIRTIAAI RSFPHDNVED LIRGGADVNC THGTLKPLHC ACMVSDADCV ELLLEKGAEV NALDGYNRTA LHYAAEKDEA CVEVLLEYGA NPNALDGNRD TPLHWAAFKN NAECVRALLE SGASVNALDY NN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
May be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications
ASB8 | |
Polyclonal | |
Unconjugated | |
ASB8 | |
4930539L19Rik; AI788835; ankyrin repeat and SOCS box containing 8; ankyrin repeat and SOCS box protein 8; ankyrin repeat and SOCS box-containing 8; ankyrin repeat and SOCS box-containing protein 8; ASB8; ASB-8; C430011H06Rik; PP14212 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140461 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H765 | |
ASB8 | |
Recombinant protein corresponding to Human ASB8. Recombinant protein control fragment (Product #RP-102075). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction