Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASB8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988843
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8843 Supplier Bioss Supplier No. BS9132R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.

Specifications

Antigen ASB8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ASB8
Gene Accession No. Q9H765
Gene Alias 4930539L19Rik; AI788835; ankyrin repeat and SOCS box containing 8; Ankyrin repeat and SOCS box protein 8; ankyrin repeat and SOCS box-containing 8; ankyrin repeat and SOCS box-containing protein 8; Asb8; ASB-8; C430011H06Rik; PP14212
Gene Symbols ASB8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ASB8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140461
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.