Learn More
Invitrogen™ ASB9 Polyclonal Antibody

Description
Recommended positive controls: HeLaS3, HepG2, Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ASB9 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Asb9 |
| Gene Accession No. | Q96DX5 |
| Gene Alias | 1700011M07Rik; ankyrin repeat and SOCS box containing 9; ankyrin repeat and SOCS box protein 9; ankyrin repeat and SOCS box-containing 9; ankyrin repeat and SOCS box-containing protein 9; ankyrin repeat and suppressor of cytokine signaling box protein 9; Asb9; ASB-9; DKFZp564L0862; FLJ20636; MGC4954; testis tissue sperm-binding protein Li 57p |
| Gene Symbols | Asb9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.