Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551795 Supplier Invitrogen™ Supplier No. PA551795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GMDGSKPAGP RDFPGIRLLS NPLMGDAVSD WSPMHEAAIH GHQLSLRNLI SQGWAVNIIT ADHVSPLHEA CLGGHLSCVK ILLKHGAQVN GVTADWHTPL FNACV Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 72%.

Substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. Recognizes at least two forms of creatine kinase, CKB and CKMT1A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Asb9
Gene Accession No. Q96DX5
Gene Alias 1700011M07Rik; ankyrin repeat and SOCS box containing 9; ankyrin repeat and SOCS box protein 9; ankyrin repeat and SOCS box-containing 9; ankyrin repeat and SOCS box-containing protein 9; ankyrin repeat and suppressor of cytokine signaling box protein 9; Asb9; ASB-9; DKFZp564L0862; FLJ20636; MGC4954; testis tissue sperm-binding protein Li 57p
Gene Symbols Asb9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASB9. Recombinant protein control fragment (Product #RP-92183).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140462
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.