Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASB9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551811
Description
Immunogen sequence: SVQPESDLAS PIHEAARRGH VECVNSLIAY GGNIDHKISH LGTPLYLACE NQQRACVKKL LESGADVNQG KGQDSPLHAV ARTASEELAC LLMDFGADTQ AKNAEGKRPV ELVPPESPLA QLFLEREGA Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.
Substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. Recognizes at least two forms of creatine kinase, CKB and CKMT1A.
Specifications
ASB9 | |
Polyclonal | |
Unconjugated | |
Asb9 | |
1700011M07Rik; ankyrin repeat and SOCS box containing 9; ankyrin repeat and SOCS box protein 9; ankyrin repeat and SOCS box-containing 9; ankyrin repeat and SOCS box-containing protein 9; ankyrin repeat and suppressor of cytokine signaling box protein 9; Asb9; ASB-9; DKFZp564L0862; FLJ20636; MGC4954; testis tissue sperm-binding protein Li 57p | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140462 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DX5 | |
Asb9 | |
Recombinant protein corresponding to Human ASB9. Recombinant protein control fragment (Product #RP-92184). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction