Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASC/TMS1 Antibody (1C3D7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261682
Description
ASC/TMS1 Monoclonal specifically detects ASC/TMS1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ASC/TMS1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
Mouse | |
21.6 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1C3D7 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
PYCARD | |
This ASC/TMS1 Antibody (1C3D7) was developed against purified recombinant fragment of human ASC/TMS1 (AA: 1-120) expressed in E. Coli. | |
Protein G purified | |
Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity | |
29108 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction