Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASC/TMS1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178977AF405
Description
ASC/TMS1 Polyclonal specifically detects ASC/TMS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ASC/TMS1 | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Q9ULZ3 | |
PYCARD | |
This ASC/TMS1 Antibody was developed against a synthetic peptide made to an N-terminal portion of the human ASC/TMS1 protein (between residues 1-50) [Uniprot: Q9ULZ3] | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity | |
29108 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Reactivity with Rat reported in PMID 24464748 . | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction