Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASC/TMS1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177297
Description
ASC/TMS1 Polyclonal specifically detects ASC/TMS1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ASC/TMS1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
| Rabbit | |
| 22 kDa | |
| 0.1 mg | |
| Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity | |
| 29108 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 ug/mL, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/mL | |
| BAA87339 | |
| PYCARD | |
| This ASC/TMS1 Antibody was developed against a 14 amino acid peptide near the carboxy terminus of human ASC/TMS1 . The immunogen is located within the last 50 amino acids of ASC. Amino Acid Sequence: RESQSYLVEDLERS | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This ASC/TMS1 Antibody can detect all human isoforms - isoform 1 (195aa, 22kDa), isoform 2 (176aa, 20kDa), and isoform 3 (135aa, 15kDa), | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction