Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100419
Description
ASC1 Polyclonal specifically detects ASC1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| ASC1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| activating signal cointegrator 1, ASC-1, HsT17391, thyroid hormone receptor interactor 4, thyroid receptor interacting protein 4, Thyroid receptor-interacting protein 4, TR-interacting protein 4, TRIP-4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:10000, Immunoprecipitation 1:10-1:500 | |
| NP_057297.2 | |
| TRIP4 | |
| A synthetic peptide mapping to a region between residues 75 and 125 of human ASC1 using the numbering given in entry NP_057297.2 (GeneID 9325. | |
| 100 μL | |
| Apoptosis, Cancer, DNA Repair, Tumor Suppressors | |
| 9325 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction