Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590403 Supplier Invitrogen™ Supplier No. PA590403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIDDESDYFA SDSNQWLSKL ERETLQKREE ELRELRHASR LSKKVTIDFA GRKILEEENS LAEYHSRLDE TIQAIANGTL NQPLTKLDRS SEEPLGVLVN PNMYQSPPQW VDHTGAASQK KAFRSSGFGL EFNSFQHQLR IQDQEFQEGF DGGWCLSVHQ PWASLLVRGI KRVEGRSWYT PHRGRLWIAA TAKKPSPQEV SELQATYRLL RGKDVEFPND YPSGCLLGCV DLIDCLSQKQ FKEQFPDISQ ESDSPFVFIC KNPQEMVVKF PIKGNPKIWK LDSKIHQGAK KGLMKQNKAV; Positive Samples: A-549, HeLa, LO2, Mouse liver, Mouse testis, Mouse brain, Rat testis, Rat brain; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, cytosol, microtubule organizing center.

Transcription coactivator of nuclear receptors which functions in conjunction with CBP-p300 and SRC-1 and may play an important role in establishing distinct coactivator complexes under different cellular conditions. Plays a pivotal role in the transactivation of NF-kappa-B, SRF and AP1. Acts as a mediator of transrepression between nuclear receptor and either AP1 or NF-kappa-B. Plays a role in androgen receptor transactivation and in testicular function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASC1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Trip4
Gene Accession No. Q15650, Q9QXN3
Gene Alias 4930558E03Rik; Activating signal cointegrator 1; Asc1; ASC-1; BB191711; HsT17391; SMABF1; thyroid hormone receptor interactor 4; thyroid receptor-interacting protein 4; transcription coactivator Asc1; TR-interacting protein 4; TRIP4; TRIP-4; ZC2HC5; zinc finger, C2HC5-type
Gene Symbols Trip4
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315769, 56404, 9325
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.