Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASC2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501557560
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7560 Supplier Bethyl Laboratories, Inc Supplier No. A300410A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat

Nuclear receptor coactivator that directly binds nuclear receptors and stimulates the transcriptional activities in a hormone-dependent fashion. Coactivates expression in an agonist- and AF2-dependent manner. Involved in the coactivation of different nuclear receptors, such as for steroids (GR and ERs), retinoids (RARs and RXRs), thyroid hormone (TRs), vitamin D3 (VDR) and prostanoids (PPARs). Probably functions as a general coactivator, rather than just a nuclear receptor coactivator. May also be involved in the coactivation of the NF-kappa-B pathway. May coactivate expression via a remodeling of chromatin and its interaction with histone acetyltransferase proteins.

Specifications

Antigen ASC2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NCOA6
Gene Accession No. Q14686
Gene Alias activating signal cointegrator 2, activating signal cointegrator-2, AIB3, amplified in breast cancer protein 3, amplified in breast cancer-3 protein, ASC2, ASC-2, cancer-amplified transcriptional coactivator ASC-2, KIAA0181, NRC, NRC RAP250, nuclear receptor coactivator RAP250, nuclear receptor-activating protein, 250 kDa, peroxisome proliferator-activated receptor interacting protein, peroxisome proliferator-activated receptor-interacting protein, PPAR-interacting protein, PRIP, RAP250, thyroid hormone receptor binding protein, thyroid hormone receptor-binding protein, TRBP
Gene Symbols NCOA6
Host Species Rabbit
Immunogen Between 1050 and 1100
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23054
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.