Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASCC1 Antibody (OTI6G5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271879F
Description
ASCC1 Monoclonal specifically detects ASCC1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ASCC1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ASCC1 | |
Full length recombinant protein of human ASCC1 (NP_057031) produced in HEK293T cells. | |
0.1 mL | |
51008.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI6G5 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
activating signal cointegrator 1 complex subunit 1, ASC-1 complex subunit p50, ASC1p50, CGI-18, Em:AC022392.3, RP11-150D20.4, Trip4 complex subunit p50 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction