Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    ASCC2 Antibody (OTI1E8), DyLight 550, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272273R
Description
ASCC2 Monoclonal specifically detects ASCC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| ASCC2 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ASCC2 | |
| Full length recombinant protein of human ASCC2 (NP_115580) produced in HEK293T cells. | |
| 0.1 mL | |
| 84164.0 | |
| Store at 4°C in the dark. | 
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1E8 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| activating signal cointegrator 1 complex subunit 2, ASC 1 complex subunit P100, ASC-1 complex subunit p100, ASC1p100, DKFZp586O0223, FLJ21588, Trip4 complex subunit p100 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            