Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASCC2 Antibody (OTI1E8), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272273PECY55
Description
ASCC2 Monoclonal specifically detects ASCC2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
ASCC2 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
ASCC2 | |
Full length recombinant protein of human ASCC2 (NP_115580) produced in HEK293T cells. | |
0.1 mL | |
84164.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI1E8 | |
Flow Cytometry | |
activating signal cointegrator 1 complex subunit 2, ASC 1 complex subunit P100, ASC-1 complex subunit p100, ASC1p100, DKFZp586O0223, FLJ21588, Trip4 complex subunit p100 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction