Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASCC2 Monoclonal Antibody (OTI2D3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503246S


Description
ASCC2 Monoclonal antibody specifically detects ASCC2 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
ASCC2 | |
Monoclonal | |
0.8 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96EK2 | |
Mouse | |
Affinity chromatography | |
RUO | |
112885 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Immunocytochemistry, Western Blot | |
OTI2D3 | |
Unconjugated | |
PHF21B | |
1700011I11Rik; 2610034L15Rik; activating signal cointegrator 1 complex subunit 2; AI482016; ASC-1 complex subunit P100; Asc1p100; ASCC2; AW046480; p100; RGD1561422; trip4 complex subunit p100 | |
Full length human recombinant protein of human PHF21B produced in HEK293 cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction