Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASCC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550623
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550623 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550623 Supplier Invitrogen™ Supplier No. PA550623
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ASCC2 protein.

Plays a role in DNA damage repair as component of the ASCC complex. Recruits ASCC3 and ALKBH3 to sites of DNA damage by binding to polyubiquitinated proteins that have 'Lys-63'-linked polyubiquitin chains (PubMed:29144457). Part of the ASC-1 complex that enhances NF-kappa-B, SRF and AP1 transactivation (PubMed:12077347). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCC2
Applications Western Blot
Classification Polyclonal
Concentration 2.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ASCC2
Gene Accession No. Q91WR3, Q9H1I8
Gene Alias 1700011I11Rik; 2610034L15Rik; activating signal cointegrator 1 complex subunit 2; AI482016; ASC-1 complex subunit P100; Asc1p100; ASCC2; AW046480; p100; RGD1561422; trip4 complex subunit p100
Gene Symbols ASCC2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human activating signal cointegrator 1 complex subunit 2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75452, 84164
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.