Learn More
ASCC2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304020AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Enhances NF-kappa-B, SRF and AP1 transactivation.Specifications
ASCC2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q91WR3, Q9H1I8 | |
Ascc2 | |
Between 707 and 757 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ASCC2 | |
ASC1p100, p100, ASC-1 complex subunit P100, trip4 complex subunit p100 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
75452, 84164 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.