Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASCC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556794 Supplier Thermo Scientific Supplier No. PA556794
Add to Cart
Add to Cart

ASCC3 Polyclonal Antibody

ASCC3 Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCC3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASCC3
Gene Accession No. Q8N3C0
Gene Alias Activating signal cointegrator 1 complex subunit 3; ASC-1 complex subunit P200; ASC1p200; ASCC3; B630009I04Rik; BC023451; D430001L07Rik; D630041L21; dJ121G13.4; dJ467N11.1; Helic1; Helicase, ATP binding 1; RNA helicase family; RNAH; Trip4 complex subunit p200
Gene Symbols ASCC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASCC3. Recombinant protein control fragment (Product #RP-94679).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10973
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.