Learn More
ASCC3 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304014A

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that belongs to a family of helicases that are involved in the ATP-dependent unwinding of nucleic acid duplexes. The encoded protein is the largest subunit of the activating signal cointegrator 1 complex that is involved in DNA repair and resistance to alkylation damage. Alternate splicing results in multiple transcript variants.Specifications
ASCC3 | |
Polyclonal | |
Unconjugated | |
ASCC3 | |
ASC-1 complex subunit P200, ASC1p200, dJ121G13.4, dJ467N11.1, HELIC1, helicase, ATP binding 1, RNA helicase family, RNAH, trip4 complex subunit p200 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10973, 77987 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
E9PZJ8, Q8N3C0 | |
Ascc3 | |
Between 175 and 225 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.