Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma Ascentis™ Express HPLC Column, 2.7 μm

Catalog No. M50514U Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
2.1 mm
3.0 mm
4.6 mm
Length (Metric):
5 cm
10 cm
15 cm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
M50514U 4.6 mm 10 cm
M50502U 2.1 mm 5 cm
M50503U 2.1 mm 10 cm
M50504U 2.1 mm 15 cm
M50505U 3.0 mm 5 cm
M50506U 3.0 mm 10 cm
M50510U 3.0 mm 15 cm
M50513U 4.6 mm 5 cm
M50515U 4.6 mm 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. M50514U Supplier MilliporeSigma Supplier No. 50514U
Add to Cart
Add to Cart

HPLC column for chromatography applications.

  • This product is a Design for Sustainability (DfS) developed Greener Alternative product, which belongs to one of the four categories of greener alternatives.
  • Has a phosphorylcholinezwitterionic stationary phase covalently attached to Fused-Core™ particle.
  • The permanent and hydrophiliczwitterion functionality makes the column suitable for Hydrophilic Interaction Liquid Chromatography (HILIC).
  • Weak electrostatic interactions between charged analytes and the neutral zwitterionic stationary phase results in a unique selectivity, and especially suitable for analytes that are poorly retained on reversed phase columns.
  • Can be used as a tool to change the selectivity or to improve peak resolution for polar and hydrophilic compounds such as carbohydrates, metabolites, acids and bases, organic and inorganic ions, metal complexes, amino acids, peptides, and protein digests.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 10 cm
Particle Size 2.7 μm
Pore Size 160 Å
Phase ZIC-cHILIC Phase
Material Stainless Steel
Technique HPLC, LC/MS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.