Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Ascites Conditioning Reagent for Melon™ Gel Monoclonal IgG Purification Kit PROMO

Catalog No. PIA45219PM
Click to view available options
:
Thermo Scientific™ Ascites Conditioning Reagent for Melon™

Catalog No. PIA45219PM

Supplier: Thermo Scientific™ 45219

Purify mouse monoclonal antibodies from serum, ascites or hybridoma cell culture supernatant with this high-yield, mild monoclonal IgG purification kit.

The Melon Gel Monoclonal IgG Purification Kit consists of a large package size of Melon Gel Resin and an optimized protocol for easy and mild purification of monoclonal IgG antibodies from hybridoma culture supernatant or ascites fluid. Unlike typical bind-and-elute affinity methods, Melon Gel Purification is based on the binding of non-IgG proteins from a prepared supernatant or ascites sample, thereby allowing the pure IgG to be recovered in the flow-through (non-bound) fraction. Antibodies are recovered ready for use in assays or subsequent conjugation and other modification steps. There is no need to neutralize or desalt to mitigate the effects of harsh elution buffers. Melon Gel Purification saves time compared to classical Protein A or Protein G methods without compromising the quality of the antibody.

Highlights:

  • Fast – purify antibodies in half the time of required for Protein A or G

  • Efficient – recovery and purity is often better than Protein A or G purification

  • Robust – effective for subclasses of IgG that bind poorly to Protein A or G

  • Gentle – antibodies retain activity because they are not exposed to denaturing conditions

  • Reusable – support can be regenerated and used for up to 6 purifications

  • Compatible – amine-free buffer does not require removal or neutralization

  • Flexible – accommodates a broad range of sample volumes and processing vessels

Includes:

Melon Gel Resin, purification buffer and resin regenerant

Specifications

Product Type Ascites Conditioning Reagent
Format Kit
Reagent Type Conditioning Reagent
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Quantity 5 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.