Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASCIZ Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179016R
Description
ASCIZ Polyclonal antibody specifically detects ASCIZ in Human samples. It is validated for Western BlotSpecifications
ASCIZ | |
Polyclonal | |
Western Blot | |
ASCIZ, ATM INteracting protein, ATM interactor, ATM/ATR-substrate CHK2-interacting zinc finger protein, ATM/ATR-Substrate Chk2-Interacting Zn++-finger protein, DKFZp779K1455, FLJ50270, FLJ61620, FLJ76795, KIAA0431, zinc finger protein 822, ZNF822 | |
The immunogen recognized by this antibody maps to a region between residue 773 and 823 of human ATM Interactor using the numbering given in entry NP_056066.2 (GeneID 23300). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23300 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction