Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASCL1/Mash1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299418MFV500
Description
ASCL1/Mash1 Polyclonal antibody specifically detects ASCL1/Mash1 in Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
ASCL1/Mash1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
achaete-scute complex (Drosophila) homolog-like 1, achaete-scute complex homolog 1 (Drosophila), achaete-scute complex-like 1 (Drosophila), achaete-scute homolog 1, ASH-1, ASH1MASH1, BHLHA46, bHLHa46achaete-scute complex-like 1, Class A basic helix-loop-helix protein 46, hASH1, HASH1achaete scute protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ASCL1/Mash1 | |
0.1 mL | |
Neuroscience | |
429 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction