Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASCL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585754 Supplier Invitrogen™ Supplier No. PA585754
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%). Positive Control: SH-SY5Y, SH-SY5Y nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ASCL1 (also known as ASH1) is a basic helix-loop-helix transcription factor that is required for early development of the nervous system. Expressed in fetal brain, ASCL1 is essential for the proper development of autonomic neurons and for the survival of subsets of autonomic neurons. ASCL1 interaction with MEF-2A may regulate the expression of specific genes that are critical for the formation of distinct neuronal circuits within the central nervous system. The high level of ASCL1 expression in neuroendocrine tumors, such as medullary thyroid cancer, small cell lung cancer and lung cancer with neuroendocrine features may provide a useful marker for cancers with neuroendocrine features. Mapping to human chromosome 12, the ASCL1 gene contains a trinucleotide repeat region, making this locus a candidate for inherited disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Ascl1
Gene Accession No. P19359, P50553, Q02067
Gene Alias achaete scute protein; achaete-scute complex homolog 1; achaete-scute complex homolog 1 (Drosophila); achaete-scute complex homolog-like 1; achaete-scute complex-like 1; achaete-scute complex-like protein 1; achaete-scute family bHLH transcription factor 1; achaete-scute homolog 1; AI225900; Ascl1; ASH1; ASH-1; bHLHa46; Class A basic helix-loop-helix protein 46; HASH1; KMT2H; mammalian achaete scute homolog 1; Mash1; mASH-1
Gene Symbols Ascl1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of mouse ASCL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17172, 429, 64186
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.