Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASCL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541489 Supplier Invitrogen™ Supplier No. PA541489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RPQAVRPSAP RGPPGTTPVA ASPSRASSSP GRGGSSEPGS PRSAYSSDDS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 79%; Pig: 100%; Rat: 93%.

This gene is a member of the basic helix-loop-helix (BHLH) family of transcription factors. It activates transcription by binding to the E box (5'-CANNTG-3'). Dimerization with other BHLH proteins is required for efficient DNA binding. Involved in the determination of the neuronal precursors in the peripheral nervous system and the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCL2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ASCL2
Gene Accession No. Q99929
Gene Alias 2410083I15Rik; achaete scute-like protein 2; achaete-scute complex homolog 2; achaete-scute complex homolog 2 (Drosophila); achaete-scute complex homolog-like 2; achaete-scute complex-like 2; achaete-scute complex-like protein 2; achaete-scute family bHLH transcription factor 2; achaete-scute homolog 2; Ascl2; ASH2; ASH-2; bHLH transcription factor; Mash2; BHLHA45; Class A basic helix-loop-helix protein 45; H M bHLHa45; HASH2; long transient receptor potential-related channel 5; mammalian achaete scute homolog 2; mammalian achaete/scute homologue 2; MASH2; mASH-2
Gene Symbols ASCL2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ASCL2 (aa 121-170).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 430
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.