Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASCL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547852
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547852 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547852 Supplier Invitrogen™ Supplier No. PA547852
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

This gene is a member of the basic helix-loop-helix (BHLH) family of transcription factors. It activates transcription by binding to the E box (5'-CANNTG-3'). Dimerization with other BHLH proteins is required for efficient DNA binding. Involved in the determination of the neuronal precursors in the peripheral nervous system and the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ASCL2
Gene Accession No. Q99929
Gene Alias 2410083I15Rik; achaete scute-like protein 2; achaete-scute complex homolog 2; achaete-scute complex homolog 2 (Drosophila); achaete-scute complex homolog-like 2; achaete-scute complex-like 2; achaete-scute complex-like protein 2; achaete-scute family bHLH transcription factor 2; achaete-scute homolog 2; Ascl2; ASH2; ASH-2; bHLH transcription factor; Mash2; BHLHA45; Class A basic helix-loop-helix protein 45; H M bHLHa45; HASH2; long transient receptor potential-related channel 5; mammalian achaete scute homolog 2; mammalian achaete/scute homologue 2; MASH2; mASH-2
Gene Symbols ASCL2
Host Species Sheep
Immunogen E. coli-derived recombinant human ASCL2/Mash2 Met1-Ala49.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 430
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
What was the pH of the buffer used for validating the ASCL2 Polyclonal Antibody (Cat. No. PA5-47852)?

The pH of the buffer used for validating the ASCL2 Polyclonal Antibody (Cat. No. PA5-47852) was pH 9-9.5.

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.