Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASCL3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP168871
Description
ASCL3 Polyclonal specifically detects ASCL3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
ASCL3 | |
Polyclonal | |
Western Blot 0.1 ug/ml, Immunohistochemistry 0.1-0.3ug/ml, Immunohistochemistry-Paraffin, Peptide ELISA detection limit 1:8000 | |
NP_064435.1 | |
ASCL3 | |
Peptide with sequence C-HLPEDYLEKRLSK corresponding to internal region according to Uniprot Mouse NP_064435.1. | |
0.1 mg | |
Primary | |
Expected from sequence similarity: Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
achaete-scute complex homolog 3 (Drosophila), ASH-3, bHLH transcription factor Sgn-1 (Salivary Glands 1), bHLH transcriptional regulator Sgn-1, BHLHA42, bHLHa42achaete-scute homolog 3, Class A basic helix-loop-helix protein 42, hASH3, HASH3achaete-scute complex (Drosophila) homolog-like 3, SGN1 | |
Goat | |
Affinity Purified | |
RUO | |
56676 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction