missing translation for 'onlineSavingsMsg'
Learn More

ASCL3 Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
SDP

Catalog No. 501684300
Encompass_Preferred
missing translation for 'changeView'
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 50-168-4300 Supplier OriGene missing translation for 'supplierNo' TA811864
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ASCL3 is a protein coding gene, transcriptional repressor, and inhibits myogenesis.

Specifications

Antigen ASCL3
Applications Western Blot
Classification Monoclonal
Clone OTI1C5
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ASCL3
Gene Accession No. Q9NQ33
Gene Alias HASH3, SGN1, bHLHa42
Gene Symbols ASCL3
Host Species Mouse
Immunogen Full length human recombit protein of human Ascl3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56676
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.