Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASCL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569011
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569011 Supplier Invitrogen™ Supplier No. PA569011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

ASCL3 is a protein coding gene, transcriptional repressor, and inhibits myogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCL3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ASCL3
Gene Accession No. Q9NQ33
Gene Alias achaete-scute complex homolog 3; achaete-scute complex homolog 3 (Drosophila); achaete-scute complex homolog-like 3; achaete-scute family bHLH transcription factor 3; achaete-scute homolog 3; Ascl3; ASH-3; ASH3A; bHLH transcription factor Sgn-1 (Salivary Glands 1); bHLH transcriptional regulator Sgn-1; bHLHa42; class A basic helix-loop-helix protein 42; HASH3; ICRFP703B1614Q5.4; ICRFP703N2430Q5.4; mASH3; mASH-3; Sgn1
Gene Symbols ASCL3
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human ASCL3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56676
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.