Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASCL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543650 Supplier Invitrogen™ Supplier No. PA543650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: METRKPAERL ALPYSLRTAP LGVPGTLPGL PRRDPLRVAL RLDAACWEWA Sequence homology: Human: 100%.

ASCL4, a basic helix-loop-helix transcription factor, is essential for the determination of cell fate and the development and differentiation of numerous tissues. It could be a transcriptional regulator involved in skin development.Basic helix-loop-helix transcription factors, such as ASCL4, are essential for the determination of cell fate and the development and differentiation of numerous tissues (Jonsson et al., 2004 [PubMed 15475265]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCL4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ASCL4
Gene Accession No. Q6XD76
Gene Alias achaete-scute complex homolog 4; achaete-scute complex-like 4; achaete-scute family bHLH transcription factor 4; achaete-scute homolog 4; Achaete-scute-like protein 4; ASCL4; ASH-4; BHLHA44; Class A basic helix-loop-helix protein 44; class II bHLH protein ASCL4; HASH4
Gene Symbols ASCL4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ASCL4 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121549
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.