Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ascrinvacumab Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542071
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Ascrinvacumab has been used in trials studying the treatment and basic science of Neoplasms, Advanced Solid Tumors, Carcinoma, Hepatocellular, and Malignant Pleural Mesothelioma.
Specifications
Ascrinvacumab | |
Recombinant Monoclonal | |
Unconjugated | |
PF-03446962 | |
Human ACVRL1/SKR3/TGF-B/ TSR-I. | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
5.91 mg/mL | |
PBS with no preservative; pH 7.5 | |
Human | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction