Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASCT2 Recombinant Rabbit Monoclonal Antibody (HL1617)
GREENER_CHOICE

Catalog No. PIMA547112
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547112 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547112 Supplier Invitrogen™ Supplier No. MA547112
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

SLC1A5 (Solute carrier family 1 (neutral amino acid transporter), member 5) is a member of the Na(+)-dependent amino acid transporter superfamily. It has a broad substrate specificity, a preference for zwitterionic amino acids, and a sodium-dependence. It accepts as substrates all neutral amino acids, including glutamine, asparagine, and branched-chain and aromatic amino acids, and excludes methylated amino acids, anionic amino acids, and cationic amino acids. It acts as a cell surface receptor for feline endogenous virus RD114, baboon M7 endogenous virus and type D simian retroviruses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCT2
Applications Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone HL1617
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SLC1A5
Gene Accession No. Q15758
Gene Alias AAAT; ASC-like Na(+)-dependent neutral amino acid transporter ASCT2; Asct2; ATB(0); ATB0; ATBO; baboon M7 virus receptor; CAZ-associated structural protein; H4-ASCT2; H4-system ASC-like transporter; insulin-activated amino acid transporter; M7V1; M7VS1; neutral amino acid transporter B; neutral amino acid transporter B(0); R16; RD114 virus receptor; RD114/simian type D retrovirus receptor; RDR; RDRC; SLC1A5; Slc1a7; sodium-dependent neutral amino acid transporter ASCT2; sodium-dependent neutral amino acid transporter type 2; solute carrier family 1 (neutral amino acid transporter), member 5; solute carrier family 1 member 5; solute carrier family 1, member 7
Gene Symbols SLC1A5
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SLC1A5. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6510
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.