Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144558
Description
ASE1 Polyclonal Antibody for Western Blot, ELISA
Required for anaphase spindle elongation.
Specifications
ASE1 | |
Polyclonal | |
Unconjugated | |
ASE1 | |
Anaphase spindle elongation protein ASE1 YOR058C YOR29-09 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
854223 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P50275 | |
ASE1 | |
Recombinant Saccharomyces cerevisiae (strain ATCC 204508/S288c) (Baker's yeast) ASE1 protein (43-223aa). | |
100 μL | |
Primary | |
Yeast | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction