Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Asenapine maleate

Catalog No. AC466310010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466310010 1 g
AC466312500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466310010 Supplier Thermo Scientific Chemicals Supplier No. 466310010
Add to Cart

CAS: 85650-56-2 | C21H20ClNO5, C17H16ClNO·C4H4O4 | 401.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85650-56-2
Molecular Formula C17H16ClNO·C4H4O4

Specifications

Quantity 1 g
Formula Weight 401.84
InChI Key GMDCDXMAFMEDAG-TXUHOWFZSA-N
SMILES OC(=O)\C=C/C(O)=O.CN1C[C@@H]2[C@@H](C1)C1=CC(Cl)=CC=C1OC1=CC=CC=C21
IUPAC Name (2R,6R)-9-chloro-4-methyl-13-oxa-4-azatetracyclo[12.4.0.0²,⁶.0⁷,¹²]octadeca-1(18),7,9,11,14,16-hexaene; (2Z)-but-2-enedioic acid
Molecular Weight (g/mol) 401.84
Chemical Name or Material Asenapine maleate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 288-064-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.