Learn More
ASF Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302053A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Zebrafish, X. tropicalis, Bovine, Pig and Orangutan
This gene encodes a member of the arginine/serine-rich splicing factor protein family, and functions in both constitutive and alternative pre-mRNA splicing. The protein binds to pre-mRNA transcripts and components of the spliceosome, and can either activate or repress splicing depending on the location of the pre-mRNA binding site. The protein′s ability to activate splicing is regulated by phosphorylation and interactions with other splicing factor associated proteins. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
ASF | |
Polyclonal | |
Unconjugated | |
SRSF1 | |
ASF, SF2, SF2p33, SFRS1, SRp30a, ASF-1, SR splicing factor 1, alternative-splicing factor 1, pre-mRNA-splicing factor SF2, P33 subunit, splicing factor 2, splicing factor, arginine/serine-rich 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6426 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q07955 | |
SRSF1 | |
Between 198 and 248 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.