Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASF/SF2 Mouse anti-Human, Mouse, Rat, Clone: 1A6C5, Proteintech
SDP

Catalog No. 501737088 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7088 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7088 Supplier Proteintech Group Inc Supplier No. 666711IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody can recognize all the 3 isoforms (28 kDa,33 kDa and 22 kDa) of SFRS1.

UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.

Specifications

Antigen ASF/SF2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1A6C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SRSF1
Gene Accession No. Q07955, Q6PDM2
Gene Alias Alternative splicing factor 1, ASF, ASF 1, ASF/SF2, SF2, SF2p33, SFRS1, SRp30a, SRSF1
Gene Symbols SRSF1
Host Species Mouse
Immunogen ASF/SF2 Fusion Protein Ag3626
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110809, 6426, 689890
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.