Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASF/SF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ASF/SF2 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8354
![]() ![]() |
Proteintech Group Inc
129292AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody can recognize all the 3 isoforms (28 kDa,33 kDa and 22 kDa) of SFRS1.
UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.Specifications
ASF/SF2 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q07955, Q6PDM2 | |
110809, 6426, 689890 | |
ASF/SF2 Fusion Protein Ag3626 | |
Primary | |
-20°C | |
SRSF1 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Alternative splicing factor 1, ASF, ASF 1, ASF/SF2, SF2, SF2p33, SFRS1, SRp30a, SRSF1 | |
SRSF1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title