Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASF1A Mouse anti-Human, Clone:2A5F8, Abnova™

Catalog No. 89334041 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-041 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-041 Supplier Abnova Corporation Supplier No. MAB17575
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human ASF1A.

This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The protein is a key component of a histone donor complex that functions in nucleosome assembly. It interacts with histones H3 and H4, and functions together with a chromatin assembly factor during DNA replication and repair. [provided by RefSeq]

Specifications

Antigen ASF1A
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2A5F8
Conjugate Unconjugated
Description ASF1 anti-silencing function 1 homolog A (S. cerevisiae)
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
Immunohistochemistry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene ASF1A
Gene Alias CGI-98, CIA, DKFZp547E2110, HSPC146
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 1-204 of human ASF1A from E. coli.
Quantity 100 μg
Gene ID (Entrez) 25842
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.