Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASF1A Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549898

Catalog No. PA549898


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total ASF1A protein.

This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The protein is a key component of a histone donor complex that functions in nucleosome assembly. It interacts with histones H3 and H4, and functions together with a chromatin assembly factor during DNA replication and repair.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ASF1A
Polyclonal
Unconjugated
ASF1A
2310079C17Rik; anti-silencing function 1A histone chaperone; anti-silencing function protein 1 homolog A; ASF1 anti-silencing function 1 homolog A; ASF1A; C79052; CCG1-interacting factor A; CGI-98; CIA; DKFZP547E2110; hAsf1; hAsf1a; hCIA; histone chaperone ASF1A; HSPC146; RP3-329L24.1
Rabbit
Antigen Affinity Chromatography
RUO
25842, 66403
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9CQE6, Q9Y294
ASF1A
Synthesized peptide derived from internal of human ASF1A.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.