Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASF1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581145
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581145 Supplier Invitrogen™ Supplier No. PA581145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ASF1A.

This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The protein is a key component of a histone donor complex that functions in nucleosome assembly. It interacts with histones H3 and H4, and functions together with a chromatin assembly factor during DNA replication and repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASF1A
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ASF1A
Gene Accession No. Q9Y294
Gene Alias 2310079C17Rik; anti-silencing function 1A histone chaperone; anti-silencing function protein 1 homolog A; ASF1 anti-silencing function 1 homolog A; ASF1A; C79052; CCG1-interacting factor A; CGI-98; CIA; DKFZP547E2110; hAsf1; hAsf1a; hCIA; histone chaperone ASF1A; HSPC146; RP3-329L24.1
Gene Symbols ASF1A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the human ASF1A.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25842
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.