Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASF1b Antibody (002), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290278APC
Description
ASF1b Monoclonal antibody specifically detects ASF1b in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ASF1b | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
002 | |
Immunohistochemistry-Paraffin | |
ASF1 silencing function 1 homolog B (S. cerevisiae), CCG1-interacting factor A-II, CIA-II, FLJ10604, hAsf1, hAsf1b, hCIA-II, histone chaperone ASF1B, silencing function protein 1 homolog B | |
Rabbit immunized with purified, recombinant Human ASF1b (Uniprot#: Q9NVP2, Met1-Ile202) | |
0.1 mL | |
Cancer, DNA replication Transcription Translation and Splicing | |
55723 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction