Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASF1b Antibody (1D4), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242676G
Description
ASF1b Monoclonal specifically detects ASF1b in Human samples. It is validated for Western Blot, ELISA.Specifications
ASF1b | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ASF1B | |
Recombinant human ASF1B (1-202aa) purified from E. coli | |
100 μL | |
Cancer, DNA replication Transcription Translation and Splicing | |
55723 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, ELISA | |
1D4 | |
Western Blot, ELISA | |
ASF1 silencing function 1 homolog B (S. cerevisiae), CCG1-interacting factor A-II, CIA-II, FLJ10604, hAsf1, hAsf1b, hCIA-II, histone chaperone ASF1B, silencing function protein 1 homolog B | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction