Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    ASF1b Antibody (1D4), mFluor Violet 610 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242676MFV610
Description
ASF1b Monoclonal antibody specifically detects ASF1b in Human samples. It is validated for Western Blot, ELISASpecifications
| ASF1b | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
| Western Blot, ELISA | |
| 1D4 | |
| Western Blot, ELISA | |
| ASF1 silencing function 1 homolog B (S. cerevisiae), CCG1-interacting factor A-II, CIA-II, FLJ10604, hAsf1, hAsf1b, hCIA-II, histone chaperone ASF1B, silencing function protein 1 homolog B | |
| Recombinant human ASF1B (1-202aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, DNA replication Transcription Translation and Splicing | |
| 55723 | |
| Store at 4C in the dark. | |
| IgG2b κ | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            