Learn More
ASF1b Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP300286AF750
Catalog No. NB065337
Only null left
Add to Cart
Description
Description
ASF1b Polyclonal specifically detects ASF1b in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
ASF1b
Polyclonal
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
ASF1 silencing function 1 homolog B (S. cerevisiae), CCG1-interacting factor A-II, CIA-II, FLJ10604, hAsf1, hAsf1b, hCIA-II, histone chaperone ASF1B, silencing function protein 1 homolog B
Produced in rabbits immunized with purified, recombinant ASF1b (Q9NVP2; Met1-Ile202).
0.1 mL
Cancer, DNA replication Transcription Translation and Splicing
55723
Store at 4°C in the dark.
IgG
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 750
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.